siRNA Details
| Gene Symbol: C7orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004323.1 | NP_001004323.1 | 3 | 679 | ||
| GenBank mRNA | BC031966.1 | - | 3 | 679 | 4839025 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7orf61| Gene Symbol: C7orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004323.1 | NP_001004323.1 | 3 | 679 | ||
| GenBank mRNA | BC031966.1 | - | 3 | 679 | 4839025 | |