siRNA Details
| Gene Symbol: ZNF624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020787.3 | NP_065838.2 | 4 | 306 | ||
| XM_006721562.3 | XP_006721625.1 | 4 | 487 | |||
| XM_011523970.2 | XP_011522272.1 | 4 | 1657 | |||
| GenBank mRNA | AK131401.1 | - | 4 | 297 | ||
| DA318803.1 | - | 4 | 281 | |||
| DA808501.1 | - | 4 | 306 | |||

ZNF624
Genomic Map