siRNA Details
| Gene Symbol: SHFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006304.1 | NP_006295.1 | 3 | 385 | ||
| GenBank mRNA | AK094899.1 | - | 2 | 2659 | ||
| AK291070.1 | - | 3 | 365 | |||
| BC032782.1 | - | 3 | 345 | 5166837 | ||
| U41515.1 | - | 3 | 385 | |||

SHFM1
Genomic Map