siRNA Details
| Gene Symbol: ARID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307939.1 | NP_001294868.1 | 9 | 2026 | ||
| NM_006465.3 | NP_006456.1 | 9 | 2023 | |||
| GenBank mRNA | AB208830.1 | - | 8 | 2070 | ||
| AF116846.1 | - | 9 | 1955 | |||
| BC041792.1 | - | 9 | 1954 | 5297774 | ||
| BC045545.1 | - | 9 | 1997 | |||

ARID3B
Genomic Map