siRNA Details
| Gene Symbol: CACYBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007214.1 | NP_001007215.1 | 6 | 1454 | ||
| NM_014412.2 | NP_055227.1 | 6 | 1639 | |||
| XM_017001046.1 | XP_016856535.1 | 6 | 1586 | |||
| GenBank mRNA | AF314752.1 | - | 6 | 1338 | ||
| AK093425.1 | - | 6 | 1426 | |||
| AL035305.1 | - | 6 | 1331 | |||
| BC005975.1 | - | 6 | 1440 | 4102572 | ||
| BC067823.1 | - | 6 | 1321 | 5260416 | ||
| BC078151.1 | - | 6 | 1304 | 6065872 | ||

CACYBP
Genomic Map