siRNA Details
| Gene Symbol: TMEM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021259.2 | NP_067082.2 | 10 | 1792 | ||
| GenBank mRNA | AB045292.1 | - | 10 | 1749 | ||
| AK024725.1 | - | 7 | 1091 | |||
| AK091823.1 | - | 10 | 1570 | |||
| AL546643.3 | - | 6 | 847 | |||
| BC021557.1 | - | 10 | 1683 | 4899167 | ||

TMEM8A
Genomic Map