siRNA Details
| Gene Symbol: CCDC121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142683.2 | NP_001136155.1 | 2 | 2661 | ||
| NM_024584.4 | NP_078860.2 | 2 | 2260 | |||
| XM_005264560.3 | XP_005264617.1 | 2 | 2780 | |||
| GenBank mRNA | AK023708.1 | - | 1 | 1756 | ||
| AK124337.1 | - | 1 | 3245 | |||
| AK125354.1 | - | 2 | 2241 | |||
| BC066936.1 | - | 2 | 2213 | 4375206 | ||
| DB554665.1 | - | 1 | 352 | |||

CCDC121
Genomic Map