siRNA Details
| Gene Symbol: AQP4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026908.1 | 7 | 629 | |||
| GenBank mRNA | AK055069.1 | - | 7 | 629 | ||
| BC105127.1 | - | 3 | 298 | |||
| BC112070.1 | - | 3 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AQP4-AS1| Gene Symbol: AQP4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026908.1 | 7 | 629 | |||
| GenBank mRNA | AK055069.1 | - | 7 | 629 | ||
| BC105127.1 | - | 3 | 298 | |||
| BC112070.1 | - | 3 | 298 | |||