siRNA Details
| Gene Symbol: PIGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184728.2 | NP_001171657.1 | 9 | 1313 | ||
| NM_001184729.2 | NP_001171658.1 | 9 | 1280 | |||
| NM_001184730.2 | NP_001171659.1 | 8 | 1175 | |||
| NM_015937.5 | NP_057021.2 | 10 | 1481 | |||
| NR_047691.1 | 10 | 1441 | ||||
| NR_047692.1 | 9 | 1384 | ||||
| NR_047693.1 | 9 | 1380 | ||||
| NR_047694.1 | 9 | 1303 | ||||
| NR_047695.1 | 7 | 1074 | ||||
| XM_005260430.2 | XP_005260487.1 | 7 | 952 | |||
| XM_005260432.2 | XP_005260489.1 | 9 | 1275 | |||
| GenBank mRNA | AB057724.1 | - | 10 | 1391 | ||
| AF132940.1 | - | 10 | 1385 | |||
| AK075469.1 | - | 10 | 1405 | |||
| AK225517.1 | - | 9 | 1210 | |||
| AK293294.1 | - | 9 | 1304 | |||
| AK293686.1 | - | 9 | 1226 | |||
| AK296063.1 | - | 9 | 1310 | |||
| AK296139.1 | - | 9 | 1236 | |||
| AK297830.1 | - | 5 | 960 | |||
| AK299013.1 | - | 7 | 1000 | |||
| AK302093.1 | - | 8 | 1133 | |||
| AK316358.1 | - | 10 | 1367 | |||
| AL121742.1 | - | 10 | 1384 | |||
| AY358588.1 | - | 10 | 1388 | |||
| BC015022.1 | - | 10 | 1410 | 3921680 | ||
| BC110892.1 | - | 10 | 1389 | |||
| BC136827.1 | - | 10 | 1419 | 9020818 | ||
| BC136828.1 | - | 10 | 1419 | 9020819 | ||
| BX537612.1 | - | 7 | 2618 | |||

PIGT
Genomic Map