siRNA Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020062.3 | NP_064446.2 | 8 | 1440 | ||
| GenBank mRNA | AB044777.1 | - | 7 | 1332 | ||
| AB044786.1 | - | 6 | 1001 | |||
| AB052777.1 | - | 8 | 1484 | |||
| AF249267.3 | - | 8 | 1385 | |||
| AK298304.1 | - | 6 | 1363 | |||
| BC001402.2 | - | 7 | 1211 | |||
| BC017446.1 | - | 5 | 1034 | |||
| BC028349.2 | - | 3 | 635 | |||
| BC052306.1 | - | 8 | 1485 | 6576325 | ||

SLC2A4RG
Genomic Map