siRNA Details
| Gene Symbol: IFT43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102564.1 | NP_001096034.1 | 7, 8 | 472 | ||
| NM_052873.2 | NP_443105.2 | 6, 7 | 487 | |||
| NR_045664.1 | 5, 6 | 392 | ||||
| NR_045665.1 | 7, 8 | 472 | ||||
| GenBank mRNA | AK056735.1 | - | 7, 8 | 783 | ||
| AK302944.1 | - | 7, 8 | 455 | |||
| BC010436.1 | - | 6, 7 | 458 | 3608220 | ||
| BI821833.1 | - | 5, 6 | 368 | |||
| CB853974.1 | - | 7, 8 | 433 | |||
| DB497730.1 | - | 7, 8 | 455 | |||

IFT43
Genomic Map