siRNA Details
| Gene Symbol: NFAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318323.1 | NP_001305252.1 | 5 | 2771 | ||
| NM_145912.6 | NP_666017.1 | 6 | 2884 | |||
| XM_005261374.4 | XP_005261431.1 | 8 | 3511 | |||
| GenBank mRNA | AK074094.1 | - | 5 | 2476 | ||
| AK097971.1 | - | 1 | 1837 | |||
| BC038241.1 | - | 6 | 2879 | 5180790 | ||

NFAM1
Genomic Map