siRNA Details
| Gene Symbol: UBE3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014671.2 | NP_055486.2 | 1, 2 | 386 | ||
| XM_005249564.4 | XP_005249621.1 | 1, 2 | 395 | |||
| GenBank mRNA | AK295138.1 | - | 1, 2 | 66 | ||
| BC014029.2 | - | 2, 3 | 146 | 4541634 | ||
| D13635.1 | - | 1, 2 | 377 | |||
| DB454915.1 | - | 1, 2 | 386 | |||

UBE3C
Genomic Map