siRNA Details
| Gene Symbol: NSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271665.1 | NP_001258594.1 | 5 | 1004 | ||
| NM_014886.4 | NP_055701.1 | 6 | 1197 | |||
| NR_073403.1 | 7 | 1306 | ||||
| XM_011543098.1 | XP_011541400.1 | 6 | 875 | |||
| XM_017008952.1 | XP_016864441.1 | 5 | 682 | |||
| GenBank mRNA | AF077615.1 | - | 6 | 911 | ||
| AF372458.1 | - | 6 | 922 | |||
| AJ012409.1 | - | 6 | 909 | |||
| BC005288.1 | - | 6 | 940 | 3683720 | ||
| CB052084.1 | - | 3 | 369 | |||

NSA2
Genomic Map