siRNA Details
| Gene Symbol: OSTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267817.1 | NP_001254746.1 | 3 | 657 | ||
| NM_001267818.1 | NP_001254747.1 | 5 | 929 | |||
| NM_021227.3 | NP_067050.1 | 4 | 855 | |||
| GenBank mRNA | AF068297.1 | - | 4 | 818 | ||
| AF161425.1 | - | 4 | 839 | |||
| AF343342.1 | - | 4 | 784 | |||
| BC016321.1 | - | 4 | 843 | 4075646 | ||
| BC054857.1 | - | 4 | 845 | 4093030 | ||

OSTC
Genomic Map