siRNA Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278505.1 | NP_001265434.1 | 7 | 920 | ||
| NM_001322249.1 | NP_001309178.1 | 8 | 1147 | |||
| NM_174931.3 | NP_777591.3 | 9 | 1189 | |||
| XM_017003748.1 | XP_016859237.1 | 9 | 1090 | |||
| GenBank mRNA | AK095667.1 | - | 7 | 853 | ||
| AK289504.1 | - | 7 | 835 | |||
| BM285380.1 | - | 9 | 1189 | |||
| BX647651.1 | - | 3 | 481 | |||

GPATCH11
Genomic Map