siRNA Details
Gene Symbol: SIX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175875.4 | NP_787071.2 | 1 | 1009 | ||
GenBank mRNA | AK074826.1 | - | 1 | 62 | ||
BU859227.1 | - | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SIX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175875.4 | NP_787071.2 | 1 | 1009 | ||
GenBank mRNA | AK074826.1 | - | 1 | 62 | ||
BU859227.1 | - | 423 |