siRNA Details
| Gene Symbol: AMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000479.3 | NP_000470.2 | 5 | 1676 | ||
| GenBank mRNA | BC049194.1 | - | 5 | 1479 | 5168299 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMH| Gene Symbol: AMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000479.3 | NP_000470.2 | 5 | 1676 | ||
| GenBank mRNA | BC049194.1 | - | 5 | 1479 | 5168299 | |