siRNA Details
| Gene Symbol: STXBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178509.5 | NP_848604.3 | 9 | 927 | ||
| XM_005257187.4 | XP_005257244.1 | 9 | 969 | |||
| XM_006721797.3 | XP_006721860.1 | 9 | 967 | |||
| XM_006721798.3 | XP_006721861.1 | 9 | 958 | |||
| XM_017024410.1 | XP_016879899.1 | 10 | 1018 | |||
| XM_017024411.1 | XP_016879900.1 | 10 | 1010 | |||
| XM_017024412.1 | XP_016879901.1 | 9 | 961 | |||
| XM_017024413.1 | XP_016879902.1 | 7 | 682 | |||
| XM_017024414.1 | XP_016879903.1 | 10 | 1017 | |||
| XM_017024415.1 | XP_016879904.1 | 10 | 1017 | |||
| GenBank mRNA | AK122865.1 | - | 9 | 927 | ||
| BC041485.2 | - | 8 | 770 | |||
| BC125116.1 | - | 9 | 916 | 40118892 | ||

STXBP4
Genomic Map