siRNA Details
Gene Symbol: ZNF428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182498.3 | NP_872304.2 | 3 | 1334 | ||
GenBank mRNA | BC045799.1 | - | 3 | 1075 | 4813427 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182498.3 | NP_872304.2 | 3 | 1334 | ||
GenBank mRNA | BC045799.1 | - | 3 | 1075 | 4813427 |