siRNA Details
| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530167.2 | XP_011528469.1 | 3 | 1806 | ||
| GenBank mRNA | AK126430.1 | - | 3 | 1117 | ||
| BC118575.1 | - | 1 | 1630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf34| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530167.2 | XP_011528469.1 | 3 | 1806 | ||
| GenBank mRNA | AK126430.1 | - | 3 | 1117 | ||
| BC118575.1 | - | 1 | 1630 | |||