siRNA Details
| Gene Symbol: UBALD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182565.3 | NP_872371.1 | 2 | 458 | ||
| GenBank mRNA | BC035511.2 | - | 2 | 296 | 5091979 | |
| BC060859.1 | - | 2 | 461 | 30333976 | ||
| CT004974.1 | - | 2 | 295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBALD2| Gene Symbol: UBALD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182565.3 | NP_872371.1 | 2 | 458 | ||
| GenBank mRNA | BC035511.2 | - | 2 | 296 | 5091979 | |
| BC060859.1 | - | 2 | 461 | 30333976 | ||
| CT004974.1 | - | 2 | 295 | |||