siRNA Details
| Gene Symbol: GDF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182828.3 | NP_878248.2 | 1 | 859 | ||
| GenBank mRNA | AB158468.1 | - | 1 | 341 | ||
| AF522369.1 | - | 1 | 859 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDF7| Gene Symbol: GDF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182828.3 | NP_878248.2 | 1 | 859 | ||
| GenBank mRNA | AB158468.1 | - | 1 | 341 | ||
| AF522369.1 | - | 1 | 859 | |||