siRNA Details
| Gene Symbol: MORN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145450.1 | NP_001138922.1 | 5 | 487 | ||
| GenBank mRNA | AY367766.1 | - | 5 | 487 | ||
| BC095472.1 | - | 3 | 297 | 30561658 | ||
| BC102032.1 | - | 5 | 384 | 40033539 | ||
| BC102034.1 | - | 5 | 384 | 40033540 | ||
| BC102035.1 | - | 5 | 384 | 40033541 | ||

MORN2
Genomic Map