siRNA Details
| Gene Symbol: LRRC56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198075.3 | NP_932341.1 | 6 | 795 | ||
| XM_005252775.2 | XP_005252832.1 | 6 | 1619 | |||
| XM_005252776.2 | XP_005252833.1 | 6 | 1619 | |||
| XM_006718132.2 | XP_006718195.1 | 6 | 1694 | |||
| XM_006718133.2 | XP_006718196.1 | 5 | 502 | |||
| XM_011519875.2 | XP_011518177.1 | 7 | 1034 | |||
| XM_011519876.1 | XP_011518178.1 | 7 | 1690 | |||
| XM_011519877.2 | XP_011518179.1 | 6 | 771 | |||
| XM_017017167.1 | XP_016872656.1 | 7 | 1109 | |||
| XM_017017168.1 | XP_016872657.1 | 7 | 1348 | |||
| GenBank mRNA | AK024495.1 | - | 5 | 2262 | ||
| BC016439.1 | - | 4 | 329 | 3447051 | ||
| BC035937.1 | - | 6 | 781 | 4816105 | ||

LRRC56
Genomic Map