siRNA Details
| Gene Symbol: SVBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199342.3 | NP_955374.1 | 2, 3 | 352 | ||
| XM_017001226.1 | XP_016856715.1 | 2, 3 | 355 | |||
| GenBank mRNA | AK291356.1 | - | 2, 3 | 247 | ||
| BC029427.1 | - | 2, 3 | 216 | 4809776 | ||
| BQ712089.1 | - | 2, 3 | 352 | |||
 Back To Top
 Back To Top
 SVBP
SVBP Genomic Map
Genomic Map Back To Top
 Back To Top