siRNA Details
| Gene Symbol: NDUFAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199069.1 | NP_951032.1 | 5 | 1254 | ||
| NM_199070.1 | NP_951033.1 | 5 | 817 | |||
| NM_199073.1 | NP_951047.1 | 5 | 888 | |||
| NM_199074.1 | NP_951056.1 | 5 | 769 | |||

NDUFAF3
Genomic Map