siRNA Details
| Gene Symbol: FAM102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035254.2 | NP_001030331.1 | 11 | 3686 | ||
| NM_203305.2 | NP_976050.1 | 8 | 3168 | |||
| GenBank mRNA | AK058004.1 | - | 4 | 1363 | ||
| AK074108.1 | - | 5 | 4143 | |||
| AL049365.1 | - | 1 | 2070 | |||
| BC047949.1 | - | 8 | 3171 | |||
| BC142952.1 | - | 6 | 2977 | |||

FAM102A
Genomic Map