siRNA Details
| Gene Symbol: LOC389199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203423.2 | NP_981968.1 | 1 | 1247 | ||
| GenBank mRNA | BC031673.1 | - | 1 | 1247 | 5166037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC389199| Gene Symbol: LOC389199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203423.2 | NP_981968.1 | 1 | 1247 | ||
| GenBank mRNA | BC031673.1 | - | 1 | 1247 | 5166037 | |