siRNA Details
| Gene Symbol: KRTAP26-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203405.1 | NP_981950.1 | 1 | 315 | ||
| GenBank mRNA | AB096936.1 | - | 1 | 76 | ||
| AM941740.1 | - | 1 | 98 | |||
| BC057825.1 | - | 1 | 316 | 30331793 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP26-1| Gene Symbol: KRTAP26-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203405.1 | NP_981950.1 | 1 | 315 | ||
| GenBank mRNA | AB096936.1 | - | 1 | 76 | ||
| AM941740.1 | - | 1 | 98 | |||
| BC057825.1 | - | 1 | 316 | 30331793 | ||