siRNA Details
| Gene Symbol: CRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249103.3 | XP_005249160.1 | 7 | 2508 | ||
| XM_017010856.1 | XP_016866345.1 | 6 | 2376 | |||
| XM_017010857.1 | XP_016866346.1 | 5 | 2304 | |||
| GenBank mRNA | AY555742.1 | - | 7 | 595 | ||
| AY555743.1 | - | 7 | 595 | |||

CRIP3
Genomic Map