siRNA Details
| Gene Symbol: MAMDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206920.2 | NP_996803.2 | 24, 25 | 3133 | ||
| GenBank mRNA | AL137659.1 | - | 22, 23 | 3585 | ||
| AL834531.1 | - | 6, 7 | 1510 | |||
| AY358419.1 | - | 24, 25 | 3133 | |||
| BF339007.1 | - | 5, 6 | 518 | |||

MAMDC4
Genomic Map