siRNA Details
| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 4 | 4616 | |||
| NR_027687.1 | 3 | 4586 | ||||
| GenBank mRNA | AK130777.1 | - | 2 | 1002 | ||
| AK310631.1 | - | 2 | 764 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00176| Gene Symbol: LINC00176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027686.1 | 4 | 4616 | |||
| NR_027687.1 | 3 | 4586 | ||||
| GenBank mRNA | AK130777.1 | - | 2 | 1002 | ||
| AK310631.1 | - | 2 | 764 | |||