siRNA Details
| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021192.2 | NP_067015.2 | 2 | 854 | ||
| GenBank mRNA | AK309681.1 | - | 2 | 178 | ||
| BC109394.1 | - | 3 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD11| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021192.2 | NP_067015.2 | 2 | 854 | ||
| GenBank mRNA | AK309681.1 | - | 2 | 178 | ||
| BC109394.1 | - | 3 | 356 | |||