siRNA Details
| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021192.2 | NP_067015.2 | 1 | 100 | ||
| GenBank mRNA | BC109394.1 | - | 1 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD11| Gene Symbol: HOXD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021192.2 | NP_067015.2 | 1 | 100 | ||
| GenBank mRNA | BC109394.1 | - | 1 | 100 | ||