siRNA Details
| Gene Symbol: ASB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212556.2 | NP_997721.2 | 6 | 1324 | ||
| GenBank mRNA | AK123854.1 | - | 5 | 796 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASB18| Gene Symbol: ASB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212556.2 | NP_997721.2 | 6 | 1324 | ||
| GenBank mRNA | AK123854.1 | - | 5 | 796 | ||