siRNA Details
| Gene Symbol: ASTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002036.3 | NP_001002036.3 | 5 | 418 | ||
| XM_011511205.2 | XP_011509507.1 | 4 | 1233 | |||
| XM_011511206.2 | XP_011509508.1 | 5 | 1456 | |||
| XM_011511207.2 | XP_011509509.1 | 4 | 1234 | |||
| XM_011511208.2 | XP_011509510.1 | 4 | 1234 | |||
| GenBank mRNA | AJ537600.2 | - | 5 | 418 | ||
| BC107126.1 | - | 5 | 417 | |||
| BC107127.1 | - | 5 | 418 | 40023980 | ||

ASTL
Genomic Map