siRNA Details
| Gene Symbol: SLC25A47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207117.2 | NP_997000.2 | 6 | 798 | ||
| GenBank mRNA | AB073385.1 | - | 7 | 722 | ||
| AK092001.1 | - | 6 | 802 | |||
| AY569438.1 | - | 6 | 798 | |||
| BC137253.1 | - | 6 | 778 | 9021250 | ||
| BC137254.1 | - | 6 | 778 | 9021251 | ||
| BC144549.1 | - | 6 | 839 | |||
| BX248260.1 | - | 6 | 787 | |||

SLC25A47
Genomic Map