siRNA Details
| Gene Symbol: SIGLEC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213602.2 | NP_998767.1 | 6 | 1357 | ||
| GenBank mRNA | AK092746.1 | - | 3 | 908 | ||
| AK095432.1 | - | 7 | 1061 | |||
| AK172835.1 | - | 6 | 1357 | |||
| AY971516.1 | - | 6 | 1346 | |||

SIGLEC15
Genomic Map