siRNA Details
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024337.3 | NP_077313.3 | 4 | 1749 | ||
XM_017009845.1 | XP_016865334.1 | 3 | 1875 | |||
GenBank mRNA | AY335938.1 | - | 4 | 1752 | ||
BC029160.1 | - | 4 | 1451 |
