siRNA Details
| Gene Symbol: GDF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001557.2 | NP_001001557.1 | 2 | 1940 | ||
| GenBank mRNA | AJ537424.1 | - | 2 | 1940 | ||
| BC043222.2 | - | 1 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDF6| Gene Symbol: GDF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001557.2 | NP_001001557.1 | 2 | 1940 | ||
| GenBank mRNA | AJ537424.1 | - | 2 | 1940 | ||
| BC043222.2 | - | 1 | 1094 | |||