siRNA Details
| Gene Symbol: SPOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001664.2 | NP_001001664.1 | 10, 11 | 1443 | ||
| XM_005263655.4 | XP_005263712.1 | 11, 12 | 1406 | |||
| XM_005263656.4 | XP_005263713.1 | 11, 12 | 1335 | |||
| XM_005263657.4 | XP_005263714.1 | 10, 11 | 1248 | |||
| GenBank mRNA | BC071613.1 | - | 10, 11 | 1422 | 5275316 | |
| BX538143.1 | - | 1, 2 | 62 | |||

SPOPL
Genomic Map