siRNA Details
| Gene Symbol: ZNF470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001668.3 | NP_001001668.3 | 3 | 708 | ||
| XM_017026801.1 | XP_016882290.1 | 3 | 708 | |||
| GenBank mRNA | AY484591.1 | - | 3 | 224 | ||
| BC136762.1 | - | 3 | 220 | 9020752 | ||
| BC142627.1 | - | 3 | 703 | |||
| BC150188.1 | - | 3 | 542 | |||

ZNF470
Genomic Map