siRNA Details
| Gene Symbol: ZNF470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001668.3 | NP_001001668.3 | 6 | 1810 | ||
| XM_017026801.1 | XP_016882290.1 | 7 | 1884 | |||
| GenBank mRNA | AK129686.1 | - | 1 | 574 | ||
| AY484591.1 | - | 6 | 1326 | |||
| BC136762.1 | - | 6 | 1322 | 9020752 | ||
| BX641145.1 | - | 1 | 773 | |||

ZNF470
Genomic Map