siRNA Details
| Gene Symbol: ARHGEF37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001669.2 | NP_001001669.2 | 13 | 4221 | ||
| XM_005268447.4 | XP_005268504.1 | 13 | 4192 | |||
| XM_005268448.1 | XP_005268505.1 | 12 | 4095 | |||
| XM_006714784.3 | XP_006714847.1 | 14 | 4560 | |||
| XM_011537642.2 | XP_011535944.1 | 13 | 4243 | |||
| XM_017009460.1 | XP_016864949.1 | 12 | 4334 | |||
| XM_017009461.1 | XP_016864950.1 | 11 | 3984 | |||
| GenBank mRNA | AK023814.1 | - | 1 | 3260 | ||
| AK123597.1 | - | 5 | 2861 | |||

ARHGEF37
Genomic Map