siRNA Details
| Gene Symbol: DDI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001711.2 | NP_001001711.1 | 1 | 521 | ||
| GenBank mRNA | AF429971.1 | - | 1 | 517 | ||
| AK093336.1 | - | 1 | 501 | |||
| BC021172.2 | - | 1 | 462 | 4822028 | ||
| BC021710.1 | - | 1 | 465 | 4826837 | ||
| BC022017.2 | - | 1 | 488 | 4825124 | ||
| BC022018.1 | - | 1 | 509 | 4825958 | ||

DDI1
Genomic Map