siRNA Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002919.2 | NP_001002919.2 | 6 | 878 | ||
| XM_005264689.4 | XP_005264746.1 | 6 | 891 | |||
| XM_006711879.3 | XP_006711942.1 | 6 | 889 | |||
| XM_011510342.2 | XP_011508644.1 | 5 | 819 | |||
| GenBank mRNA | AY358517.1 | - | 6 | 344 | ||
| BC033490.1 | - | 6 | 752 | |||

FAM150B
Genomic Map