siRNA Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004127.2 | NP_001004127.2 | 3 | 881 | ||
| GenBank mRNA | AK025456.1 | - | 3 | 854 | ||
| AK128503.1 | - | 2 | 3786 | |||
| AK296747.1 | - | 3 | 871 | |||
| BC010857.2 | - | 1 | 463 | |||
| BC073862.1 | - | 1 | 37 | |||
| BC111022.1 | - | 3 | 854 | |||
| BC142998.1 | - | 3 | 880 | 8860455 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183142, NM_001243161, BC061469, AK046475, AK134312, and others.

ALG11
Genomic Map