siRNA Details
| Gene Symbol: C1orf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004303.4 | NP_001004303.3 | 20 | 3426 | ||
| GenBank mRNA | BX648439.1 | - | 20 | 3104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf168| Gene Symbol: C1orf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004303.4 | NP_001004303.3 | 20 | 3426 | ||
| GenBank mRNA | BX648439.1 | - | 20 | 3104 | ||