siRNA Details
| Gene Symbol: RTP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004312.2 | NP_001004312.2 | 2 | 1164 | ||
| XM_017006301.1 | XP_016861790.1 | 5 | 1358 | |||
| XM_017006302.1 | XP_016861791.1 | 5 | 1309 | |||
| GenBank mRNA | BC068081.1 | - | 2 | 1164 | 6212901 | |

RTP2
Genomic Map